Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centaurin beta 2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP224528

 View more versions of this product

Catalog No. NBP224528


Only null left
Add to Cart

Description

Description

Centaurin beta 2 Polyclonal specifically detects Centaurin beta 2 in Human, Mouse, Rat, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Centaurin beta 2
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Arf GAP with coiled coil, ANK repeat and PH domains 2, ArfGAP with coiled-coil, ankyrin repeat and PH domains 2, centaurin, beta 2, Centaurin-beta-2, CENTB2centaurin-beta-2, CNT-B2, KIAA0041arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2
Rabbit
Affinity Purified
RUO
23527
Human, Mouse, Rat, Canine, Equine, Primate
IgG
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 0.25-0.5 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Q15057
ACAP2
A portion of amino acids 350-400 of human ACAP2 was used as the immunogen for the antibody.
0.1 mg
Primary
The amino acid sequence used as immunogen for the antibody is 100% homologous in mouse and 87% homologous in human and monkey.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only