Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centaurin beta 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | Centaurin beta 2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.25-0.5 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224528SS
![]() |
Novus Biologicals
NBP224528SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224528
![]() |
Novus Biologicals
NBP224528 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
Centaurin beta 2 Polyclonal specifically detects Centaurin beta 2 in Human, Mouse, Rat, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Centaurin beta 2 | |
Western Blot 0.25-0.5 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat, Canine, Equine, Primate | |
Q15057 | |
23527 | |
A portion of amino acids 350-400 of human ACAP2 was used as the immunogen for the antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
Arf GAP with coiled coil, ANK repeat and PH domains 2, ArfGAP with coiled-coil, ankyrin repeat and PH domains 2, centaurin, beta 2, Centaurin-beta-2, CENTB2centaurin-beta-2, CNT-B2, KIAA0041arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2 | |
ACAP2 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen for the antibody is 100% homologous in mouse and 87% homologous in human and monkey. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title