Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centaurin beta 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001321 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001321 Supplier Novus Biologicals Supplier No. NB1001321
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Centaurin beta 2 Polyclonal specifically detects Centaurin beta 2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen Centaurin beta 2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_036419.
Gene Alias Arf GAP with coiled coil, ANK repeat and PH domains 2, ArfGAP with coiled-coil, ankyrin repeat and PH domains 2, centaurin, beta 2, Centaurin-beta-2, CENTB2centaurin-beta-2, CNT-B2, KIAA0041arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2
Gene Symbols ACAP2
Host Species Goat
Immunogen Peptide with sequence C-EKLNRFQQDSQKF corresponding to C-Terminus according to NP_036419.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23527
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.