Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centaurin beta 2 Rabbit anti-Human, Mouse, Rat, Clone: S05-8H3, Novus Biologicals™
SDP

Catalog No. NB159287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159287 25 μL
NB188376 100 ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159287 Supplier Novus Biologicals Supplier No. NBP31501125UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Centaurin beta 2 Monoclonal antibody specifically detects Centaurin beta 2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Centaurin beta 2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S05-8H3
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias Arf GAP with coiled coil, ANK repeat and PH domains 2, ArfGAP with coiled-coil, ankyrin repeat and PH domains 2, centaurin, beta 2, Centaurin-beta-2, CENTB2centaurin-beta-2, CNT-B2, KIAA0041arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2
Host Species Rabbit
Immunogen A synthetic peptide of human Centaurin beta 2 (Uniprot#: Q15057)
Purification Method Affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23527
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.