Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENTB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095530 100 μL
NB095531 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095530 Supplier Novus Biologicals Supplier No. NBP303562100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CENTB1 Polyclonal specifically detects CENTB1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CENTB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Arf GAP with coiled coil, ANK repeat and PH domains 1, arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 1, ArfGAP with coiled-coil, ankyrin repeat and PH domains 1, centaurin, beta 1, Centaurin-beta-1, CENTB1cnt-b1, Cnt-b1, KIAA0050centaurin-beta-1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-230 of human CENTB1 (NP_055531.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9744
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.