Learn More
Eppendorf™ Centrifuge Bucket
Shop All Eppendorf North America Products
Click to view available options
No. of Places:
4
Description
- Buckets for: Rotor S-24-11-AT, Rotor A-4-62, Rotor A-4-81
Specifications
Specifications
| For Use With (Equipment) | 4 x 1.5mL/2mL Eppendorf tubes of S-24-11-AT rotor |
| Capacity | 4 x 1.5/2 mL |
| Container Type | Bucket |
| No. of Places | 4 |
| Quantity | 2/Pk. |