Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Centrin 3 Antibody (OTI4G5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP270409A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270409A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270409A94 Supplier Novus Biologicals Supplier No. NBP270409AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Centrin 3 Monoclonal specifically detects Centrin 3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Centrin 3
Applications Western Blot
Classification Monoclonal
Clone OTI4G5
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245
Gene Symbols CETN3
Host Species Mouse
Immunogen Full length recombinant protein of human Centrin 3 (NP_004356) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1070
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.