Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centrisart™ I centrifugal ultrafiltration unit, Sold by MilliporeSigma™ Supelco™
Click to view available options
Pore Size:
100 kDa MWCO
Specifications
Specifications
Pore Volume | 2.5 mL |
Quantity | 12 Tubes |
Surface Area | 0.79 cm2 |
Volume (Metric) | 15 mL |
Dimensions | 14 x 93 mm |
Material | Polystyrene |
Centrisart is a registered trademark of Sartorius Stedim Biotech GmbH
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction