Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Centromere protein R Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 107431AP150UL

Description
ITGB3BP is a transcription coregulator that can have both coactivator and corepressor functions. Isoform 1, but not other isoforms, is involved in the coactivation of nuclear receptors for retinoid X (RXRs) and thyroid hormone (TRs) in a ligand-dependent fashion. ITGB3BP acts as a transcriptional corepressor via its interaction with the NFKB1 NF-kappa-B subunit, possibly by interfering with the transactivation domain of NFKB1. It induces apoptosis in breast cancer cells, but not in other cancer cells, via a caspase-2 mediated pathway that involves mitochondrial membrane permeabilization but does not require other caspases. ITGB3BP may also act as an inhibitor of cyclin A-associated kinase. ITGB3BP may be involved in incorporation of newly synthesized CENPA into centromeres via its interaction with the CENPA-NAC complex.Specifications
Centromere protein R | |
Polyclonal | |
Unconjugated | |
ITGB3BP | |
Beta 3 endonexin, CENP R, CENPR, Centromere protein R, HSU37139, ITGB3BP, NRIF3, TAP20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23421 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13352 | |
ITGB3BP | |
Centromere protein R Fusion Protein Ag1204 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction