Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP131/AZ1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7144278
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1966 10 μL
50-157-1965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1966 Supplier Bethyl Laboratories, Inc Supplier No. IHC00363T
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee and Northern white-cheeked gibbon

CEP131 (Centrosomal Protein 131) is a Protein Coding gene. Diseases associated with CEP131 includeAnterior Spinal Artery Syndrome. Among its related pathways areOrganelle biogenesis and mainteceandCell Cycle, Mitotic. Gene Ontology (GO) annotations related to this gene includeprotein homodimerization activity.

Specifications

Antigen CEP131/AZ1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CEP131
Gene Accession No. Q9UPN4
Gene Alias 5-azacytidine induced 1, 5-azacytidine-induced protein 1, AZ1, AZI1, centrosomal protein 131 kDa, centrosomal protein 131kDa, KIAA1118, pre-acrosome localization protein 1, ZA1
Gene Symbols CEP131
Host Species Rabbit
Immunogen Between 1033 and 1083
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22994
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.