Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP290 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1086991A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1086991A30 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1086991A30 Supplier Novus Biologicals Supplier No. NB10086991AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEP290 Polyclonal specifically detects CEP290 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen CEP290
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BBS14Bardet-Biedl syndrome 14 protein, Cancer/testis antigen 87, centrosomal protein 290kDa, Cep290, CT87JBTS6, FLJ13615, JBTS5CTCL tumor antigen se2-2, KIAA0373FLJ21979, LCA10monoclonal 3H11 antigen, MKS4, Nephrocystin-6, NPHP6centrosomal protein of 290 kDa, POC3, POC3 centriolar protein homolog, prostate cancer antigen T21, rd16,3H11AG, SLSN6nephrocytsin-6,3H11Ag, Tumor antigen se2-2
Gene Symbols CEP290
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2429 and 2479 of human centrosomal protein 290kDa using the numbering given in entry NP_079390.3 (GeneID 80184).
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 80184
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.