Learn More
CEP290 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | CEP290 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-0931
|
Bethyl Laboratories, Inc
A301659AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells.
CEP290 activates ATF4 mediated transcription and is required for the correct localization of ciliary and phototransduction proteins in retinal photoreceptor cells; may play a role in ciliary transport processes. CEP290 is ubiquitously expressed; strongly in placenta and weakly in brain. There are two named isoforms. Defects in CEP290 are a cause of Joubert syndrome type 5 (JBTS5), Senior-Loken syndrome type 6 (SLSN6), Leber congenital amaurosis type 10 (LCA10) and Meckel syndrome type 4 (MKS4). Antibodies against CEP290 are present in sera from patients with cutaneous T cell lymphomas, but not in the healthy control population.Specifications
| CEP290 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Centrosomal protein of 290 kDa, Cep290, Bardet-Biedl syndrome 14 protein, Cancer/testis antigen 87, CT87, Nephrocystin-6, Tumor antigen se2-2, CEP290, BBS14, KIAA0373, NPHP6 | |
| CEP290 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O15078 | |
| 80184 | |
| Between 2429 and 2479 | |
| Primary | |
| 4° C | |
| CEP290 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.