Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP63 Antibody (OTI2H2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP272221A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272221A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272221A32 Supplier Novus Biologicals Supplier No. NBP272221AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEP63 Monoclonal specifically detects CEP63 in Human samples. It is validated for Western Blot.

Specifications

Antigen CEP63
Applications Western Blot
Classification Monoclonal
Clone OTI2H2
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias centrosomal protein 63kDa, centrosomal protein of 63 kDa, centrosome protein CEP63, Cep63, FLJ13386, MGC78416
Gene Symbols CEP63
Host Species Mouse
Immunogen Full length recombinant protein of human CEP63 (NP_001035859) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80254
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.