Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cepharanthine, Thermo Scientific™

Catalog No. AC468912500
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Chemical Name or Material Cepharanthine
CAS 481-49-2
Quantity 250 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C37 H38 N2 O6
Physical Form Crystalline Powder
Color White to Light-yellow to Beige
Purity ≥94%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.