Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceramide Kinase Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339735 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339735 Supplier Novus Biologicals Supplier No. NB1002911MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ceramide Kinase Polyclonal specifically detects Ceramide Kinase in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Ceramide Kinase
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ceramide kinase, dA59H18.2, DKFZp434E0211, EC 2.7.1.138, FLJ21430, FLJ23239, hCERKLipid kinase 4, KIAA1646dA59H18.3, lipid kinase LK4, LK4Acylsphingosine kinase, MGC131878
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Ceramide Kinase (between residues 50-150). [Swiss-Prot# Q8TCT0]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 64781
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.