Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceramide Kinase Antibody, Novus Biologicals™
SDP

Catalog No. NB1002911SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002911SS 0.025 mL
NB1002911 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002911SS Supplier Novus Biologicals Supplier No. NB1002911SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Ceramide Kinase Polyclonal specifically detects Ceramide Kinase in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Ceramide Kinase
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:500-1:2000
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Accession No. Q8TCT0
Gene Alias ceramide kinase, dA59H18.2, DKFZp434E0211, EC 2.7.1.138, FLJ21430, FLJ23239, hCERKLipid kinase 4, KIAA1646dA59H18.3, lipid kinase LK4, LK4Acylsphingosine kinase, MGC131878
Gene Symbols CERK
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Ceramide Kinase (between residues 50-150). [Swiss-Prot# Q8TCT0]
Molecular Weight of Antigen 65 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 64781
Test Specificity Human. Immunogen has 86% homology to mouse.
Target Species Human, Mouse, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.