Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ceramide
GREENER_CHOICE

Catalog No. p-4783323
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Ser/Thr protein phosphatase activator

A potent modulator of cell proliferation and differentiation. Activates protein phosphatase-1 (PP1) and -2A (PP2A), as well as ceramide-activated protein phosphatase (CAPP) in vitro.

Chemical Identifiers

CAS 3102-57-6
Molecular Formula C20H39NO3

Specifications

Quantity 10 mg
Formula Weight 341.53
Purity >98%
Molecular Weight (g/mol) 341.54
Chemical Name or Material Ceramide
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.