Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Ceritinib

Click to view available options
Quantity:
10 mg
50 mg
Description
Ceritinib is a highly potent anaplastic lymphoma kinase (ALK) inhibitor (IC50 = 0.2 nM). Also displays potent inhibition of insulin receptor (IR), insulin-like growth factor receptor 1 (IGF1R), serine/threonine-protein kinase STK22D and FLT3 (IC50 values are 7, 8, 23 and 60 nM, respectively). Inhibits tumor growth in Karpas299 and H2228 rat xenograft models.
Specifications
Specifications
Synonym | LDK378 |
Quantity | 10 mg |
Molecular Formula | C28H36ClN5O3S |
Formula Weight | 558.14 |
Product Type | Inhibitor |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction