Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) carbonate hydrate, REacton™, 99.999% (REO)

Catalog No. p-7059379
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 54451-25-1 | C3Ce2O9 | 460.26 g/mol

Cerium(III) carbonate hydrate is used to prepare cerium chloride and incandescent lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54451-25-1
Molecular Formula C3Ce2O9
Molecular Weight (g/mol) 460.26
MDL Number MFCD00149635
InChI Key GHLITDDQOMIBFS-UHFFFAOYSA-H
Synonym cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reo
PubChem CID 16212049
IUPAC Name dicerium(3+) tricarbonate
SMILES [Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 848°C
Physical Form Crystalline
Quantity 25 g
Merck Index 14,1996
Solubility Information Soluble in dilute mineral acids. Insoluble in water.
Formula Weight 460.27 (Anhydrous)
Percent Purity 99.999%
Grade Reacton™
Assay (REO)
Chemical Name or Material Cerium (III) carbonate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-655-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.