Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) sulfate, 99.9%, (REO=Rare Earth Oxides)

Catalog No. AC378660500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378660500 50 g Glass bottle
AC378662500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC378660500 Supplier Thermo Scientific Chemicals Supplier No. 378660500
Only null left
Add to Cart

CAS: 13454-94-9 | Ce2O12S3 | 568.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-94-9
Molecular Formula Ce2O12S3
Molecular Weight (g/mol) 568.43
MDL Number MFCD00010931
InChI Key OZECDDHOAMNMQI-UHFFFAOYSA-H
Synonym cerium sulfate, cerous sulfate, cerium iii sulfate, cerium sulphate, dicerium trisulfate, cerium sesquisulfate, unii-0vf70o14r1, sulfuric acid, cerium salt, dicerium trisulphate, sulphuric acid, cerium salt
PubChem CID 159674
IUPAC Name cerium(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+3].[Ce+3]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 g
Assay Percent Range 46.0 to 50% (Ce) (Complexometry)
Linear Formula Ce2(SO4)3
Merck Index 15, 2001
Solubility Information Solubility in water: slightly soluble
Formula Weight 568.43
Percent Purity 99.90%
Packaging Glass bottle
Chemical Name or Material Cerium(III) sulfate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-644-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.