Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cerium(III) trifluoromethanesulfonate, 98%

Catalog No. p-7048115
Click to view available options
Quantity:
5 g
25 g

CAS: 76089-77-5 | C3CeF9O9S3 | 587.31 g/mol

It is employed as intermediate for organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for organic synthesis and pharmaceuticals.

Solubility
Soluble in water.

Notes
Hygroscopic. store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76089-77-5
Molecular Formula C3CeF9O9S3
Molecular Weight (g/mol) 587.31
MDL Number MFCD02093899
InChI Key PHSMPGGNMIPKTH-UHFFFAOYSA-K
Synonym cerium iii trifluoromethanesulfonate, cerium iii triflate, trifluoromethanesulfonic acid cerium iii salt, cerium trifluoromethanesulfonate, cerous triflate, ce otf 3, 3cf3o3s.ce, acmc-209p1w, ksc491c9f
PubChem CID 2733941
IUPAC Name cerium(3+);trifluoromethanesulfonate
SMILES [Ce+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point >300°C
Density 1.7
Linear Formula (CF3SO3)3Ce
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 587.33
Percent Purity 98%
Chemical Name or Material Cerium(III) trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.