Learn More
Cerium(IV) ammonium nitrate, 98+%
Description
Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.
Solubility
Soluble in water and alcohol.
Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents and powdered metals.
Specifications
Specifications
| Quantity | 50 g |
| Linear Formula | (NH4)2Ce(NO3)6 |
| UN Number | UN1477 |
| Merck Index | 14,1992 |
| Solubility Information | Soluble in water and alcohol. |
| Formula Weight | 548.23 |
| Percent Purity | ≥98% |
| Odor | Pungent |
| Chemical Name or Material | Cerium(IV) ammonium nitrate |
Safety and Handling
| Hazard Category | H272-H290-H302-H314-H317-H335 |
| Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S. |
| EINECSNumber | 240-827-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.