Learn More
Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO)
Description
Cerium(IV) ammonium nitrate is used to offers a novel route to tartronic acid derivatives via facile oxygenation of malonates. It is also used as for oxidative addition reactions of electrophilic radicals to alkenes, enabling intermolecular and intramolecular carbon-carbon and carbon-heteroatom bond formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Orange |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Linear Formula | (NH4)2Ce(NO3)6 |
| UN Number | UN1477 |
| Merck Index | 14,1992 |
| Solubility Information | Soluble in water. |
| Formula Weight | 548.23 |
| Percent Purity | 99.5% |
| Odor | Pungent |
| Show More |
Safety and Handling
| Hazard Category | H272-H290-H302-H314-H317-H335 |
| Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: II |
| EINECSNumber | 240-827-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.