Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Certified Clean Borosilicate Glass Vials

Catalog No. p-7204500 Shop All MilliporeSigma Supelco Products
Click to view available options
No. per Pack:
6
100

Borosilicate glass vials with certificate included

  • Case includes individual certificate
  • Certified Vials have <0.010 mg/L C from TOC
  • Include dust caps
  • Cleaning Products Cosmetics
  • Environmental Flavors and Fragrances
  • Food and Beverages
  • Genteral Analytical Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Cap 24 mm
Material Silicone, PTFE
For Use With (Application) TOC Analysis
No. per Pack 100
Volume (English) 40 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.