Alfa Aesar™ Cesium acetate, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 1292914

Quantity 25g

 View more versions of this product

Catalog No. AA1292914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3396-11-0
    Formula Weight 191.95
    MDL Number MFCD00013056
    Chemical Name or Material Cesium acetate

    Specifications

    Formula Weight 191.95
    Odor Weak
    CAS 3396-11-0
    Melting Point 194°C
    TSCA TSCA
    Sensitivity Hygroscopic
    MDL Number MFCD00013056
    Physical Form Crystalline
    Quantity 25g
    Chemical Name or Material Cesium acetate
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit