Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis

Catalog No. p-222835
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle

Catalog No. AC189520500

Supplier: Thermo Scientific Chemicals 189520500

Only null left
Add to Cart

CAS: 35103-79-8 | CsH3O2 | 167.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35103-79-8
Molecular Formula CsH3O2
Molecular Weight (g/mol) 167.93
MDL Number MFCD00149664
InChI Key ABSOMGPQFXJESQ-UHFFFAOYSA-M
Synonym cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
PubChem CID 23679066
SMILES O.[OH-].[Cs+]

Specifications

Melting Point 205.0°C to 208.0°C
Color Blue-Red to Green
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.95%
Linear Formula CsOH·H2O
Merck Index 10, 1970
Solubility Information Solubility in water: soluble with exothermic reaction. Other solubilities: soluble in alcohol (exothermic)
IUPAC Name caesium(1+) hydrate hydroxide
Formula Weight 167.93
Percent Purity 99.95%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Cesium hydroxide monohydrate, 99.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.