Learn More
Cesium iodide, ultra dry, 99.998% (metals basis)
Description
Cesium iodide is used as a beam splitter in Fourier transform infrared (FT-IR) spectrometers. It is widely used as a scintillator in silicon photodiodes. It is utilized in calorimeters and detectors and also employed as phosphor in fluoroscopy equipments. It finds application in making cell windows and prisms in infrared spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Boiling Point | 1280°C |
| Melting Point | 626°C |
| Color | White |
| Physical Form | Beads |
| Quantity | 10 g |
| Merck Index | 14,2014 |
| Solubility Information | Soluble in water and ethanol. Slightly soluble in methanol. Insoluble in acetone. |
| IUPAC Name | cesium;iodide |
| Formula Weight | 259.81 |
| Percent Purity | 99.998% |
| Show More |
Safety and Handling
| Hazard Category | H315-H317-H319-H335-H361 |
| Hazard Statement | GHS H Statement H302-H315-H319-H317-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 232-145-2 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.