Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7050653
Click to view available options
Quantity:
25 g

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14644-55-4
Molecular Formula CsO3V
Molecular Weight (g/mol) 231.844
MDL Number MFCD00049812
InChI Key SZIZYHNGMIYCQE-UHFFFAOYSA-N
Synonym Cesium vanadate, meta
PubChem CID 23689378
IUPAC Name cesium;oxido(dioxo)vanadium
SMILES [O-][V](=O)=O.[Cs+]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 25 g
UN Number UN3288
Solubility Information Slightly soluble in water.
Formula Weight 231.84
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Cesium metavanadate
Hazard Category H302+H332-H370
Hazard Statement GHS H Statement
H370-H302-H332
Causes damage to organs.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P301+P312-P304+P340-P307+P311-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 238-693-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.