Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetilistat 98.0+%, TCI America™
SDP

Catalog No. C27451G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 282526-98-1
Molecular Formula C25H39NO3
Molecular Weight (g/mol) 401.591
MDL Number MFCD09839697
InChI Key MVCQKIKWYUURMU-UHFFFAOYSA-N
Synonym 2-Hexadecyloxy-6-methyl-4H-3,1-benzoxazin-4-one
PubChem CID 9952916
IUPAC Name 2-hexadecoxy-6-methyl-3,1-benzoxazin-4-one
SMILES CCCCCCCCCCCCCCCCOC1=NC2=C(C=C(C=C2)C)C(=O)O1

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 1 g
Formula Weight 401.59
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cetilistat
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.