Learn More
Cetirizine dihydrochloride, 99+%
Description
Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.
Solubility
Soluble in water, DMSO, ethanol, and methanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Merck Index | 14,2022 |
| Solubility Information | Soluble in water,DMSO,ethanol,and methanol. |
| Formula Weight | 461.82 |
| Percent Purity | ≥99% |
| Physical Form | Solid |
| Chemical Name or Material | Cetirizine dihydrochloride |
Safety and Handling
| missing translation for 'healthHazard1' | H302 |
| missing translation for 'healthHazard2' | GHS H Statement H302 Harmful if swallowed. |
| missing translation for 'healthHazard3' | P264b-P270-P301+P312-P330-P501c |
| RTECSNumber | AG0980000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.