Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CETP Antibody (ATM192), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB30558A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30558A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30558A350 Supplier Novus Biologicals Supplier No. NB300558AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CETP Monoclonal specifically detects CETP in Human, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CETP
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone ATM192
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cholesteryl ester transfer protein, cholesteryl ester transfer protein, plasma, HDLCQ10, Lipid transfer protein I
Gene Symbols CETP
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues C D(149) S G R V R T D A P D(159) of human CETP protein.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1071
Target Species Human, Bovine, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.