Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CETP Antibody (ATM192) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB300558 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300558 Supplier Novus Biologicals Supplier No. NB300558
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CETP Monoclonal specifically detects CETP in Human, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen CETP
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone ATM192
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:400, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500
Gene Accession No. P11597
Gene Alias cholesteryl ester transfer protein, cholesteryl ester transfer protein, plasma, HDLCQ10, Lipid transfer protein I
Gene Symbols CETP
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues C D(149) S G R V R T D A P D(159) of human CETP protein.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1071
Test Specificity Human and Rabbit. Not yet tested in other species.
Target Species Human, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.