Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CETP Antibody (SR1985), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32165750UL
Description
CETP Monoclonal antibody specifically detects CETP in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
CETP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
SR1985 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
cholesteryl ester transfer protein, cholesteryl ester transfer protein, plasma, HDLCQ10, Lipid transfer protein I | |
A synthesized peptide derived from human CETP (Uniprot #: P11597) | |
50 μg | |
Lipid and Metabolism | |
1071 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction