Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CETP Antibody (SR1985), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CETP |
---|---|
Clone | SR1985 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
CETP Monoclonal antibody specifically detects CETP in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
CETP | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cholesteryl ester transfer protein, cholesteryl ester transfer protein, plasma, HDLCQ10, Lipid transfer protein I | |
A synthesized peptide derived from human CETP (Uniprot #: P11597) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1985 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1071 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title