Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetuximab Antibody (2259C), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345653 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345653 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345653 Supplier Novus Biologicals Supplier No. FAB9626MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cetuximab Monoclonal specifically detects Cetuximab in Multi-species samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen Cetuximab
Applications ELISA, Flow Cytometry, Western Blot, ELISA
Classification Monoclonal
Clone 2259C
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Full length Cetuximab,
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Multi-species
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.