Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFH Monoclonal Antibody (OTI5H5), TrueMAB™, OriGene
SDP

Catalog No. 501680288
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-0288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-0288 Supplier OriGene Supplier No. TA804532
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement factor H1 (Complement factor H1, factor H1, CFH) is a plasma glycoprotein (139 kDa) and the secreted form of the protein is composed of 20 repetitive units of 60 amino acids, named short consensus repeats (SCR). Factor H1 is a central regulator of early alternative pathway activation by acting as a cofactor for factor I in the cleavage of C3b into iC3b. Factor H1 also competes with factor B for binding with C3b, inhibiting excessive alternative pathway activation. Complete (homozygous) factor H1 deficiency by genetic mutation has been rarely reported.

Specifications

Antigen CFH
Applications Western Blot
Classification Monoclonal
Clone OTI5H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CFH
Gene Accession No. P08603
Gene Alias AHUS1, AMBP1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS
Gene Symbols CFH
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 125-346 of human CFH produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3075
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.