Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Antibody (CFTR/7003R), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB157617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB157617 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB157617 Supplier Novus Biologicals Supplier No. NBP314272JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CFTR Monoclonal antibody specifically detects CFTR in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CFTR
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CFTR/7003R
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR
Host Species Rabbit
Immunogen Recombinant fragment (around aa550-850) of human CFTR protein (exact sequence is proprietary) (Uniprot: P13569 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 1080
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.