Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Mouse anti-Human, Clone-CFTR/1643, Novus Biologicals™
SDP

Catalog No. NB254509100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254509100 100 μg
NB254509200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254509100 Supplier Novus Biologicals Supplier No. NBP254509100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CFTR Monoclonal specifically detects CFTR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen CFTR
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CFTR/1643
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml
Formulation PBS with No Preservative
Gene Alias ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR
Gene Symbols CFTR
Host Species Mouse
Immunogen Recombinant human CFTR fragment (aa258-385) (exact sequence is proprietary)
Molecular Weight of Antigen 167 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 1080
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.