Learn More
Cole-Parmer™ CG-900 Cryo-Blade™ Cryogenic Grinder Processor Blade
Click to view available options
For Use With (Equipment):
Cryo-Blade
Description
- Accessories to rapidly pulverize samples from bulk to analytical preparation
- Excellent for cryogenic grinding of fruits, vegetables, seeds, stalks, roots, fungi, and flowers
- Suitable for use with the Cole-Parmer™ CG-900 Cryo-Blade™ cryogenic grinders, 61211-71 and 61211-72. Mills sold separately
Specifications
Specifications
Product Type | Processor Blade |
For Use With (Equipment) | Cryo-Blade |
Material | Metal |