Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cGAS Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB168952 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168952 20 μg
NB168951 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168952 Supplier Novus Biologicals Supplier No. NBP31693620UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

cGAS Polyclonal antibody specifically detects cGAS in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen cGAS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias C6orf150, c-GAS, cyclic GMP-AMP synthase, h-cGAS, Mab-21 domain containing 1
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S291 of mouse cGAS (NP_775562.2).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 115004
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.