Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CGP 36216 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
CGP 36216 hydrochloride is a selective GABAB antagonist. Displays activity at presynaptic receptors; ineffective at GABAB postsynaptic receptors.
Specifications
Specifications
CAS | 1781834-71-6 |
Quantity | 10 mg |
Product Type | CGP 36216 hydrochloride |
Purity | >99% |
Molecular Weight (g/mol) | 187.6 |
Inhibitors | CGP 36216 hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction