Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGP 37157
GREENER_CHOICE

Catalog No. p-4783329
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antagonist of mitochondrial Na+/Ca2+ exchange

Selective antagonist of the mitochondrial Na+-Ca2+ exchanger (IC50 = 0.4 μM).

Chemical Identifiers

CAS 75450-34-9
Molecular Formula C15H11Cl2NOS

Specifications

Quantity 10 mg
Formula Weight 324.22
Purity >99%
Molecular Weight (g/mol) 324.219
Chemical Name or Material CGP 37157
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.