Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGP 52432, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective GABAB receptor antagonist (IC50 = 85 nM).
Specifications
Quantity | 10 mg |
Formula Weight | 384.24 |
Percent Purity | >99% |
Chemical Name or Material | CGP 52432 |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction