Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGP 60474
GREENER_CHOICE

Catalog No. p-7110832
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent cdk inhibitor

CGP 60474 is a potent cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 3 - 80, 9.5, 13, 17 - 60, 200 and 220 nM for cdk2, cdk5, cdk9, cdk1, cdk4 and cdk7, respectively). Also inhibits PKCα in the low micromolar range. Displays reversible G1/S cell cycle arrest in U2-OS cells.

Specifications

Chemical Name or Material 3-[[4-[2-[(3-Chlorophenyl)amino]-4-pyrimidinyl]-2-pyridinyl]amino]-1-propanol
CAS 164658-13-3
Quantity 10 mg
Target Cyclin-dependent Kinase Inhibitors
Molecular Formula C18H18ClN5O
Formula Weight 355.82
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.