Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CGP 71683 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
CGP 71683 hydrochloride is an extremely selective, non-peptide NPY Y5 receptor antagonist. Displays > 1000-fold selectivity over Y1, Y2 and Y4 receptors; IC50 values are 1.4, 2765, 7187 and 5637 nM at cloned rat Y5, Y1, Y2 and Y4 receptors respectively. Potently inhibits NPY-induced food intake following i.p. administration in diabetic, free-feeding and fasted rats.
Specifications
Specifications
Quantity | 50 mg |
Product Type | CGP 71683 hydrochloride |
Purity | >99% |
Molecular Weight (g/mol) | 512.07 |
Inhibitors | CGP 71683 hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction