Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CGP 74514 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
CGP 74514 dihydrochloride is a potent cdk1 inhibitor (IC50 = 25 nM). Reduces Akt phosphorylation and increases mitochondrial damage in leukemia cells in vitro in combination with LY 294002 .
Specifications
Specifications
| Chemical Name or Material | rel-N2-[(1R,2S)-2-Aminocyclohexyl]-N6-(3-chlorophenyl)-9-ethyl-9H-purine-2,6-diamine dihydrochloride |
| Quantity | 10 mg |
| Target | Cyclin-dependent Kinase Inhibitors |
| Molecular Formula | C19H24ClN7.2HCl |
| Formula Weight | 458.82 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction