Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGP 78608 hydrochloride
GREENER_CHOICE

Catalog No. p-4783336
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective glycine-site NMDA antagonist

Potent and selective NMDA antagonist that acts through the glycine site (IC50 = 5 nM). Displays > 500-fold selectivity over kainate and AMPA receptors (IC50 values are 2.7 and 3 μM respectively). Also potentiates GluN1/GluN3A-mediated glycine currents (EC50 = 26.3 nM). Anticonvulsant in vivo following systemic administration.

Chemical Identifiers

CAS 1135278-54-4
Molecular Formula C11H14BrClN3O5P

Specifications

Quantity 10 mg
Formula Weight 414.58
Purity >98%
Molecular Weight (g/mol) 414.577
Chemical Name or Material CGP 78608 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.