Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGREF1 Antibody (001), PE, Novus Biologicals™
SDP

Catalog No. NB334398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334398 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334398 Supplier Novus Biologicals Supplier No. NBP290214PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CGREF1 Monoclonal antibody specifically detects CGREF1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen CGREF1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 001
Conjugate PE
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias cell growth regulator with EF hand domain protein 1, cell growth regulator with EF-hand domain 1, Cell growth regulatory gene 11 protein, CGR11cell growth regulator with EF hand domain 1, hydrophobestin
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human CGREF1 (Uniprot#: Q99674-1, Met1-Ile301)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10669
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.