Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGREF1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB063554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB063554 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB063554 Supplier Novus Biologicals Supplier No. NBP299923B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CGREF1 Polyclonal specifically detects CGREF1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CGREF1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias cell growth regulator with EF hand domain protein 1, cell growth regulator with EF-hand domain 1, Cell growth regulatory gene 11 protein, CGR11cell growth regulator with EF hand domain 1, hydrophobestin
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CGREF1 (Q99674-1; Met1-Ile301).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10669
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.