Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGRP1 Antibody (5), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB096732 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096732 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB096732 Supplier Novus Biologicals Supplier No. NBP300518AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CGRP1 Monoclonal antibody specifically detects CGRP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen CGRP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I
Host Species Mouse
Immunogen Synthetic human alpha-CGRP
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cellular Signaling, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 796
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.